missing translation for 'onlineSavingsMsg'
Learn More

Tris(8-quinolinolato)aluminum 98.0+%, TCI America™

Supplier:  TCI America T1527250G

 View more versions of this product

Catalog No. T1527250G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tris(8-quinolinolato)aluminum
418°C
C27H18AlN3O3
250 g
459.44
Crystalline Powder
2085-33-8
Yellow
MFCD00191693
tri-8-quinolinolatoaluminum, aluminum 8-hydroxyquinolinate, tri quinolin-8-yloxy alumane, aluminum tris quinolin-8-olate, tris 8-hydroxyquinolinato aluminum, aluminum, tris 8-quinolinato-n1,o8, aluminum oxinate, aluminum oxinate,, tris 8-quinolyloxy alumane, tris 8-quinolinyloxy alumane
≥98.0% (T)

Chemical Identifiers

2085-33-8
C27H18AlN3O3
MFCD00191693
tri-8-quinolinolatoaluminum, aluminum 8-hydroxyquinolinate, tri quinolin-8-yloxy alumane, aluminum tris quinolin-8-olate, tris 8-hydroxyquinolinato aluminum, aluminum, tris 8-quinolinato-n1,o8, aluminum oxinate, aluminum oxinate,, tris 8-quinolyloxy alumane, tris 8-quinolinyloxy alumane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : BD2231000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions