missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(8-quinolinolato)aluminum 98.0+%, TCI America™
$121.16 - $717.67
Chemical Identifiers
| CAS | 2085-33-8 |
|---|---|
| Molecular Formula | C27H18AlN3O3 |
| MDL Number | MFCD00191693 |
| Synonym | tri-8-quinolinolatoaluminum, aluminum 8-hydroxyquinolinate, tri quinolin-8-yloxy alumane, aluminum tris quinolin-8-olate, tris 8-hydroxyquinolinato aluminum, aluminum, tris 8-quinolinato-n1,o8, aluminum oxinate, aluminum oxinate,, tris 8-quinolyloxy alumane, tris 8-quinolinyloxy alumane |
Chemical Identifiers
| 2085-33-8 | |
| C27H18AlN3O3 | |
| MFCD00191693 | |
| tri-8-quinolinolatoaluminum, aluminum 8-hydroxyquinolinate, tri quinolin-8-yloxy alumane, aluminum tris quinolin-8-olate, tris 8-hydroxyquinolinato aluminum, aluminum, tris 8-quinolinato-n1,o8, aluminum oxinate, aluminum oxinate,, tris 8-quinolyloxy alumane, tris 8-quinolinyloxy alumane |
Specifications
| 2085-33-8 | |
| Yellow | |
| MFCD00191693 | |
| tri-8-quinolinolatoaluminum, aluminum 8-hydroxyquinolinate, tri quinolin-8-yloxy alumane, aluminum tris quinolin-8-olate, tris 8-hydroxyquinolinato aluminum, aluminum, tris 8-quinolinato-n1,o8, aluminum oxinate, aluminum oxinate,, tris 8-quinolyloxy alumane, tris 8-quinolinyloxy alumane | |
| ≥98.0% (T) | |
| Tris(8-quinolinolato)aluminum |
| 418°C | |
| C27H18AlN3O3 | |
| 5 g | |
| 459.44 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : BD2231000
TSCA : No