missing translation for 'onlineSavingsMsg'
Learn More

Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III), 99% (metals basis)

Catalog No. p-7050848
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3958203 1 g
AA3958206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3958203 Supplier Thermo Scientific Chemicals Supplier No. 03958203

CAS: 18323-96-1 | C30H30F21O6Yb | 1058.584 g/mol

It is a reactant involved in the synthesis of rare earth metal complexes for separation of lanthanide metals, conformational mobility of substituted 2-methoxychalcones under the action of lanthanide shift reagents, NMR titration on cation-π complexes of bowl-shaped polycyclic aromatic hydrocarbons, specific recognition of chiral amino alcohols. It finds its application as catalysis and synthesis, optics & glasses, healthcare & biochemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18323-96-1
Molecular Formula C30H30F21O6Yb
Molecular Weight (g/mol) 1058.584
MDL Number MFCD00044289
InChI Key KZBQCXBCJMHJOB-VNGPFPIXSA-K
Synonym tris 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato ytterbium, 6,6,7,7,8,8,8-heptafluoro-2,2-dimethyloctane-3,5-dione; ytterbium 3+, z-6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-5-oxooct-3-en-3-olate; ytterbium 3+
PubChem CID 6510275
IUPAC Name (Z)-6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-5-oxooct-3-en-3-olate;ytterbium(3+)
SMILES CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)[O-].CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)[O-].CC(C)(C)C(=CC(=O)C(C(C(F)(F)F)(F)F)(F)F)[O-].[Yb+3]

Specifications

Melting Point 108°C to 111°C
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Slowly degrades in water and soluble in hydrocarbons,alcohols,ketones,esters.
Formula Weight 1058.59
Percent Purity 99%
Assay (metals basis)
Chemical Name or Material Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato)ytterbium(III)
EINECSNumber 242-211-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only