missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-trifluoromethylphenyl)bismuth Dichloride 98.0+%, TCI America™

Catalog No. T20381G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T20381G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T20381G Supplier TCI America Supplier No. T20381G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 121882-75-5
Molecular Formula C21H12BiCl2F9
Molecular Weight (g/mol) 715.193
MDL Number MFCD06797186
InChI Key VOZWPVZQISDWJM-UHFFFAOYSA-L
Synonym Dichlorotris(4-trifluoromethylphenyl)bismuth
PubChem CID 44409351
IUPAC Name dichloro-tris[4-(trifluoromethyl)phenyl]bismuth
SMILES C1=CC(=CC=C1C(F)(F)F)[Bi](C2=CC=C(C=C2)C(F)(F)F)(C3=CC=C(C=C3)C(F)(F)F)(Cl)Cl

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3261
Formula Weight 715.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-trifluoromethylphenyl)bismuth Dichloride