missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-chlorophenyl)phosphine, 97%

CAS: 1159-54-2 | C18H12Cl3P | 365.618 g/mol

Supplier:  Thermo Scientific Chemicals A1248714

 View more versions of this product

Catalog No. AAA1248714


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tris(4-chlorophenyl)phosphine
102°C to 103°C
MFCD00013639
751458
tris 4-chlorophenyl phosphine, tri p-chlorophenyl phosphine, tris p-chlorophenyl phosphine, tris 4-chlorophenyl phosphane, tri-4-chlorophenyl phosphine, phosphine, tris 4-chlorophenyl, tri 4-chlorophenyl phosphine, acmc-20ajbt, pubchem6454
C1=CC(=CC=C1P(C2=CC=C(C=C2)Cl)C3=CC=C(C=C3)Cl)Cl
365.618
365.63
1159-54-2
C18H12Cl3P
25 g
Air sensitive
IQKSLJOIKWOGIZ-UHFFFAOYSA-N
tris(4-chlorophenyl)phosphane
70874
97%

Chemical Identifiers

1159-54-2
365.618
IQKSLJOIKWOGIZ-UHFFFAOYSA-N
70874
C1=CC(=CC=C1P(C2=CC=C(C=C2)Cl)C3=CC=C(C=C3)Cl)Cl
C18H12Cl3P
MFCD00013639
tris 4-chlorophenyl phosphine, tri p-chlorophenyl phosphine, tris p-chlorophenyl phosphine, tris 4-chlorophenyl phosphane, tri-4-chlorophenyl phosphine, phosphine, tris 4-chlorophenyl, tri 4-chlorophenyl phosphine, acmc-20ajbt, pubchem6454
tris(4-chlorophenyl)phosphane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 214-596-7

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only