missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(4-bromophenyl)amine, 98%, Thermo Scientific Chemicals
Description
Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tris(4-bromophenyl)amine was used in the synthesis of porous luminescent covalent--organic polymers (COPs)
Solubility
Soluble in toluene.
Notes
Incompatible with strong oxidizing agents. Store under room temperature.
Specifications
Specifications
| Melting Point | 141°C to 143°C |
| Quantity | 10 g |
| Solubility Information | Soluble in toluene. |
| Formula Weight | 482.01 |
| Percent Purity | 98% |
| Chemical Name or Material | Tris(4-bromophenyl)amine |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only