missing translation for 'onlineSavingsMsg'
Learn More

Tris(4-aminophenyl)methane 97.0+%, TCI America™

Catalog No. p-7137811
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T35991G 1 g
T35995G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T35991G Supplier TCI America Supplier No. T35991G

Chemical Identifiers

CAS 548-61-8
Molecular Formula C19H19N3
Molecular Weight (g/mol) 289.382
MDL Number MFCD00137820
InChI Key ADUMIBSPEHFSLA-UHFFFAOYSA-N
Synonym 4,4′,4′′-Methanetriyltrianiline, Leucopararosaniline
PubChem CID 68356
IUPAC Name 4-[bis(4-aminophenyl)methyl]aniline
SMILES C1=CC(=CC=C1C(C2=CC=C(C=C2)N)C3=CC=C(C=C3)N)N

Specifications

Melting Point 208°C
Quantity 1 g
Formula Weight 289.38
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(4-aminophenyl)methane