missing translation for 'onlineSavingsMsg'
Learn More

Tris[4-(2-thienyl)phenyl]amine 98.0+%, TCI America™

Catalog No. T30501G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30501G 1 g
T30505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30501G Supplier TCI America Supplier No. T30501G

Chemical Identifiers

CAS 142807-63-4
Molecular Formula C30H21NS3
Molecular Weight (g/mol) 491.69
MDL Number MFCD28291910
InChI Key DXPFPUHRRPAXAO-UHFFFAOYSA-N
PubChem CID 15315428
IUPAC Name 4-(thiophen-2-yl)-N,N-bis[4-(thiophen-2-yl)phenyl]aniline
SMILES S1C=CC=C1C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CS1)C1=CC=C(C=C1)C1=CC=CS1

Specifications

Melting Point 190°C
Quantity 1 g
Formula Weight 491.69
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris[4-(2-thienyl)phenyl]amine