accessibility menu, dialog, popup

UserName

SNSS

CAS RN 142807-63-4

IUPAC Name: 4-(thiophen-2-yl)-N,N-bis[4-(thiophen-2-yl)phenyl]aniline
Synonyms: 4-(thiophen-2-yl)-N,N-bis[4-(thiophen-2-yl)phenyl]aniline
Molecular Weight (g/mol): 491.69
Molecular Formula: C30H21NS3
InChi Key: DXPFPUHRRPAXAO-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=CS1)C1=CC=C(C=C1)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results