missing translation for 'onlineSavingsMsg'
Learn More

Tris[4'-(2-thienyl)-4-biphenylyl]amine 97.0+%, TCI America™

Catalog No. t3337200mg
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3337200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T3337200MG Supplier TCI America Supplier No. T3337200MG

Chemical Identifiers

CAS 1092356-36-9
Molecular Formula C48H33NS3
Molecular Weight (g/mol) 719.979
MDL Number MFCD28291947
InChI Key STWJEWBEHUGXTL-UHFFFAOYSA-N
Synonym Tris[4′C-(thiophen-2-yl)-4-biphenylyl]amine
PubChem CID 59757673
IUPAC Name 4-(4-thiophen-2-ylphenyl)-N,N-bis[4-(4-thiophen-2-ylphenyl)phenyl]aniline
SMILES C1=CSC(=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)N(C4=CC=C(C=C4)C5=CC=C(C=C5)C6=CC=CS6)C7=CC=C(C=C7)C8=CC=C(C=C8)C9=CC=CS9

Specifications

Melting Point 272°C
Color Yellow
Quantity 200 mg
Formula Weight 719.98
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Tris[4′-(2-thienyl)-4-biphenylyl]amine