accessibility menu, dialog, popup

UserName

CAS RN 1092356-36-9

IUPAC Name: 4'-(thiophen-2-yl)-N,N-bis[4'-(thiophen-2-yl)-[1,1'-biphenyl]-4-yl]-[1,1'-biphenyl]-4-amine
Synonyms: 4'-(thiophen-2-yl)-N,N-bis[4'-(thiophen-2-yl)-[1,1'-biphenyl]-4-yl]-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol): 719.98
Molecular Formula: C48H33NS3
InChi Key: STWJEWBEHUGXTL-UHFFFAOYSA-N
SMILES: S1C=CC=C1C1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CS1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=CC=CS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results