missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris[3-(trimethoxysilyl)propyl] Isocyanurate 95.0+%, TCI America™
$42.87 - $124.89
Chemical Identifiers
| CAS | 26115-70-8 |
|---|---|
| Molecular Formula | C21H45N3O12Si3 |
| Molecular Weight (g/mol) | 615.855 |
| MDL Number | MFCD00054746 |
| InChI Key | QWOVEJBDMKHZQK-UHFFFAOYSA-N |
| Synonym | Isocyanuric Acid Tris[3-(trimethoxysilyl)propyl] Ester |
| PubChem CID | 117734 |
| IUPAC Name | 1,3,5-tris(3-trimethoxysilylpropyl)-1,3,5-triazinane-2,4,6-trione |
| SMILES | CO[Si](CCCN1C(=O)N(C(=O)N(C1=O)CCC[Si](OC)(OC)OC)CCC[Si](OC)(OC)OC)(OC)OC |
Chemical Identifiers
| 26115-70-8 | |
| 615.855 | |
| QWOVEJBDMKHZQK-UHFFFAOYSA-N | |
| 117734 | |
| CO[Si](CCCN1C(=O)N(C(=O)N(C1=O)CCC[Si](OC)(OC)OC)CCC[Si](OC)(OC)OC)(OC)OC |
| C21H45N3O12Si3 | |
| MFCD00054746 | |
| Isocyanuric Acid Tris[3-(trimethoxysilyl)propyl] Ester | |
| 1,3,5-tris(3-trimethoxysilylpropyl)-1,3,5-triazinane-2,4,6-trione |
Specifications
| 26115-70-8 | |
| 236°C | |
| MFCD00054746 | |
| Isocyanuric Acid Tris[3-(trimethoxysilyl)propyl] Ester | |
| CO[Si](CCCN1C(=O)N(C(=O)N(C1=O)CCC[Si](OC)(OC)OC)CCC[Si](OC)(OC)OC)(OC)OC | |
| 615.855 | |
| 615.86 | |
| Liquid |
| Yellow | |
| C21H45N3O12Si3 | |
| 5 g | |
| QWOVEJBDMKHZQK-UHFFFAOYSA-N | |
| 1,3,5-tris(3-trimethoxysilylpropyl)-1,3,5-triazinane-2,4,6-trione | |
| 117734 | |
| ≥95.0% (GC) | |
| Tris[3-(trimethoxysilyl)propyl] Isocyanurate |
Safety and Handling
EINECSNumber : (2)-2964
RTECSNumber : XZ2025000
TSCA : Yes