missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(3-aminopropyl)amine 97.0+%, TCI America™
Supplier: TCI America T154025ML
Specifications
| Tris(3-aminopropyl)amine | |
| Yellow | |
| C9H24N4 | |
| 25 mL | |
| 3,3′,3′′-Nitrilotris(propylamine) | |
| C(CN)CN(CCCN)CCCN | |
| 188.319 | |
| 188.32 | |
| Liquid |
| 4963-47-7 | |
| 150°C | |
| MFCD00191593 | |
| 2735 | |
| QMXSDTGNCZVWTB-UHFFFAOYSA-N | |
| N',N'-bis(3-aminopropyl)propane-1,3-diamine | |
| 547030 | |
| ≥97.0% (T) |
Chemical Identifiers
| 4963-47-7 | |
| 188.319 | |
| QMXSDTGNCZVWTB-UHFFFAOYSA-N | |
| N',N'-bis(3-aminopropyl)propane-1,3-diamine |
| C9H24N4 | |
| MFCD00191593 | |
| 547030 | |
| C(CN)CN(CCCN)CCCN |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator