missing translation for 'onlineSavingsMsg'
Learn More

Tris(3-aminopropyl)amine 97.0+%, TCI America™

Catalog No. p-7136826
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15405ML 5 mL
T154025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15405ML Supplier TCI America Supplier No. T15405ML
Only null left

Chemical Identifiers

CAS 4963-47-7
Molecular Formula C9H24N4
Molecular Weight (g/mol) 188.319
MDL Number MFCD00191593
InChI Key QMXSDTGNCZVWTB-UHFFFAOYSA-N
PubChem CID 547030
IUPAC Name N',N'-bis(3-aminopropyl)propane-1,3-diamine
SMILES C(CN)CN(CCCN)CCCN

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 mL
UN Number 2735
Synonym 3,3′C,3′C′C-Nitrilotris(propylamine)
Formula Weight 188.32
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Tris(3-aminopropyl)amine
TSCA No
Recommended Storage Refrigerator