missing translation for 'onlineSavingsMsg'
Learn More

Tris[3,5-bis(trifluoromethyl)phenyl]phosphine 98.0+%, TCI America™

Catalog No. T25261G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25261G 1 g
T25265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25261G Supplier TCI America Supplier No. T25261G

Chemical Identifiers

CAS 175136-62-6
Molecular Formula C24H9F18P
Molecular Weight (g/mol) 670.281
MDL Number MFCD00042428
InChI Key ITJHLZVYLDBFOJ-UHFFFAOYSA-N
Synonym tris 3,5-bis trifluoromethyl phenyl phosphine, tris 3,5-bis trifluoromethyl phenyl phosphane, acmc-20ajat, tri 3,5-di trifluoromethyl phenyl phosphine, tris 3,5-bis-trifluoromethylphenyl phosphine, phosphine,tris 3,5-bis trifluoromethyl phenyl, tris 3,5-bis trifluoromethyl phenyl phosphine gc , yellow-brown
PubChem CID 2778032
IUPAC Name tris[3,5-bis(trifluoromethyl)phenyl]phosphane
SMILES C1=C(C=C(C=C1C(F)(F)F)P(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 101°C
Color White-Yellow
Boiling Point 130°C
Quantity 1 g
Formula Weight 670.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tris[3,5-bis(trifluoromethyl)phenyl]phosphine