missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-pyridylmethyl)amine 98.0+%, TCI America™
Supplier: TCI America T26715G
Specifications
| Tris(2-pyridylmethyl)amine | |
| 86°C | |
| C18H18N4 | |
| 5 g | |
| C(N(CC1=CC=CC=N1)CC1=CC=CC=N1)C1=CC=CC=N1 | |
| 290.37 | |
| 290.37 | |
| Crystalline Powder |
| 16858-01-8 | |
| White-Yellow | |
| MFCD14708176 | |
| VGUWFGWZSVLROP-UHFFFAOYSA-N | |
| tris[(pyridin-2-yl)methyl]amine | |
| 379259 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 16858-01-8 | |
| 290.37 | |
| VGUWFGWZSVLROP-UHFFFAOYSA-N | |
| tris[(pyridin-2-yl)methyl]amine |
| C18H18N4 | |
| MFCD14708176 | |
| 379259 | |
| C(N(CC1=CC=CC=N1)CC1=CC=CC=N1)C1=CC=CC=N1 |
Safety and Handling
TSCA : No