accessibility menu, dialog, popup

UserName

CAS RN 16858-01-8

NNNN

CAS RN 16858-01-8

IUPAC Name: tris[(pyridin-2-yl)methyl]amine
Synonyms: tris(2-pyridylmethyl)amine
Molecular Weight (g/mol): 290.37
Molecular Formula: C18H18N4
InChi Key: VGUWFGWZSVLROP-UHFFFAOYSA-N
SMILES: C(N(CC1=CC=CC=N1)CC1=CC=CC=N1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results