missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methoxyphenyl)phosphine 97.0+%, TCI America™

Catalog No. p-7137524
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
T30051G 1 g
T30055G 5 g
2 options

Catalog No. T30051G

Supplier: TCI America T30051G

Only null left

Chemical Identifiers

CAS 4731-65-1
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00014892
InChI Key IIOSDXGZLBPOHD-UHFFFAOYSA-N
Synonym tris 2-methoxyphenyl phosphine, tris o-methoxyphenyl phosphine, tris 2-methoxyphenyl phosphane, phosphine, tris methoxyphenyl, tris-2-methoxyphenyl-phosphine, tris o-anisyl phosphine, tri-2-methoxyphenyl phosphine, pubchem19707, tri 2-methoxyphenyl phosphine, ksc286k3b
PubChem CID 78464
IUPAC Name tris(2-methoxyphenyl)phosphane
SMILES COC1=CC=CC=C1P(C2=CC=CC=C2OC)C3=CC=CC=C3OC

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 1 g
Formula Weight 352.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-methoxyphenyl)phosphine