Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tris(2-methoxyphenyl)phosphine 97.0+%, TCI America™
Specifications
Specifications
| Melting Point | 204°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 352.37 |
| Percent Purity | ≥97.0% (HPLC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Tris(2-methoxyphenyl)phosphine |
Safety and Handling
| TSCA | No |