missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-methoxyphenyl)phosphine, 98%

Catalog No. p-7081198
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1109603 1 g
AAA1109606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1109603 Supplier Thermo Scientific Chemicals Supplier No. A1109603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4731-65-1 | C21H21O3P | 352.37 g/mol

Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(2-methoxyphenyl)phosphine is a phosphine ligand used for catalysis. It is used as a catalyst for allylic substitution of simple alkenes, suzuki coupling reactions, hydrogenation of quinolines and other addition reactions.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 4731-65-1
Molecular Formula C21H21O3P
Molecular Weight (g/mol) 352.37
MDL Number MFCD00014892
InChI Key IIOSDXGZLBPOHD-UHFFFAOYSA-N
Synonym tris 2-methoxyphenyl phosphine, tris o-methoxyphenyl phosphine, tris 2-methoxyphenyl phosphane, phosphine, tris methoxyphenyl, tris-2-methoxyphenyl-phosphine, tris o-anisyl phosphine, tri-2-methoxyphenyl phosphine, pubchem19707, tri 2-methoxyphenyl phosphine, ksc286k3b
PubChem CID 78464
IUPAC Name tris(2-methoxyphenyl)phosphane
SMILES COC1=CC=CC=C1P(C2=CC=CC=C2OC)C3=CC=CC=C3OC

Specifications

Melting Point 196°C to 204°C
Quantity 1 g
Beilstein 2776186
Sensitivity Air Sensitive
Solubility Information Slightly soluble in water.
Formula Weight 352.37
Percent Purity 98%
Chemical Name or Material Tris (2-methoxyphenyl)phosphine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-235-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only