missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-hydroxyethyl) Isocyanurate 98.0+%, TCI America™

Catalog No. T067425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T067425G 25 g
T0674500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T067425G Supplier TCI America Supplier No. T067425G

Chemical Identifiers

CAS 839-90-7
Molecular Formula C9H15N3O6
Molecular Weight (g/mol) 261.23
MDL Number MFCD00003549
InChI Key BPXVHIRIPLPOPT-UHFFFAOYSA-N
Synonym 1,3,5-tris 2-hydroxyethyl cyanuric acid, theic, tris 2-hydroxyethyl isocyanurate, tris hydroxyethyl cyanurate, tris 2-hydroxyethyl cyanurate, tris hydroxyethyl isocyanurate, 1,3,5-triazine-2,4,6 1h,3h,5h-trione, 1,3,5-tris 2-hydroxyethyl, 1,3,5-tris 2-hydroxyethyl-1,3,5-triazinane-2,4,6-trione, unii-n7w2mh0p2m
PubChem CID 13286
IUPAC Name tris(2-hydroxyethyl)-1,3,5-triazinane-2,4,6-trione
SMILES OCCN1C(=O)N(CCO)C(=O)N(CCO)C1=O

Specifications

Melting Point 136°C
Color White
Quantity 25 g
Formula Weight 261.23
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-hydroxyethyl) Isocyanurate
EINECSNumber (5)-1051&(5)-1058
TSCA Yes