missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-ethylhexyl) Phosphate 98.0+%, TCI America™

Catalog No. P102225ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P102225ML 25 mL
P1022500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P102225ML Supplier TCI America Supplier No. P102225ML

Chemical Identifiers

CAS 78-42-2
Molecular Formula C24H51O4P
Molecular Weight (g/mol) 434.64
MDL Number MFCD00009491
InChI Key GTVWRXDRKAHEAD-UHFFFAOYNA-N
Synonym tris 2-ethylhexyl phosphate, disflamoll tof, flexol tof, phosphoric acid, tris 2-ethylhexyl ester, kronitex tof, flexol plasticizer tof, 2-ethyl-1-hexanol phosphate, tris ethylhexyl phosphate, tri 2-ethylhexyl phosphate
PubChem CID 6537
IUPAC Name tris(2-ethylhexyl) phosphate
SMILES CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC

Specifications

Color Colorless
Boiling Point 215°C
Quantity 25 mL
Formula Weight 434.64
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-ethylhexyl) Phosphate
EINECSNumber (2)-2000
RTECSNumber MP0770000
TSCA Yes