accessibility menu, dialog, popup

UserName

H3CH3COPOOCH3CH3OH3CCH3

CAS RN 78-42-2

IUPAC Name: tris(2-ethylhexyl) phosphate
Synonyms: tris(2-ethylhexyl) phosphate
Molecular Weight (g/mol): 434.64
Molecular Formula: C24H51O4P
InChi Key: GTVWRXDRKAHEAD-UHFFFAOYNA-N
SMILES: CCCCC(CC)COP(=O)(OCC(CC)CCCC)OCC(CC)CCCC
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)