missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-cyanoethyl) Borate 98.0+%, TCI America™

Catalog No. T30245G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T30245G Supplier TCI America Supplier No. T30245G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 126755-67-7
Molecular Formula C9H12BN3O3
Molecular Weight (g/mol) 221.023
InChI Key QRJXIXWHPANYDU-UHFFFAOYSA-N
Synonym Boric Acid Tris(2-cyanoethyl) Ester
PubChem CID 90362390
IUPAC Name tris(2-cyanoethyl) borate
SMILES B(OCCC#N)(OCCC#N)OCCC#N

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
Formula Weight 221.02
Percent Purity ≥98.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Tris(2-cyanoethyl) Borate