accessibility menu, dialog, popup

UserName

CAS RN 126755-67-7

IUPAC Name: 3-{[bis(2-cyanoethoxy)boranyl]oxy}propanenitrile
Synonyms: 3-{[bis(2-cyanoethoxy)boranyl]oxy}propanenitrile
Molecular Weight (g/mol): 221.02
Molecular Formula: C9H12BN3O3
InChi Key: QRJXIXWHPANYDU-UHFFFAOYSA-N
SMILES: N#CCCOB(OCCC#N)OCCC#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results