missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-chloroethyl) Phosphate 93.0+%, TCI America™
$46.60 - $145.40
Chemical Identifiers
| CAS | 115-96-8 |
|---|---|
| Molecular Formula | C6H12Cl3O4P |
| Molecular Weight (g/mol) | 285.482 |
| MDL Number | MFCD00000967 |
| InChI Key | HQUQLFOMPYWACS-UHFFFAOYSA-N |
| Synonym | tris 2-chloroethyl phosphate, celluflex, fyrol cef, disflamoll tca, celluflex cef, niax 3cf, tris chloroethyl phosphate, trichlorethyl phosphate, 2-chloroethanol phosphate |
| PubChem CID | 8295 |
| ChEBI | CHEBI:35037 |
| IUPAC Name | tris(2-chloroethyl) phosphate |
| SMILES | C(CCl)OP(=O)(OCCCl)OCCCl |
Chemical Identifiers
| 115-96-8 | |
| 285.482 | |
| HQUQLFOMPYWACS-UHFFFAOYSA-N | |
| 8295 | |
| tris(2-chloroethyl) phosphate |
| C6H12Cl3O4P | |
| MFCD00000967 | |
| tris 2-chloroethyl phosphate, celluflex, fyrol cef, disflamoll tca, celluflex cef, niax 3cf, tris chloroethyl phosphate, trichlorethyl phosphate, 2-chloroethanol phosphate | |
| CHEBI:35037 | |
| C(CCl)OP(=O)(OCCCl)OCCCl |
Specifications
| 115-96-8 | |
| 146°C | |
| MFCD00000967 | |
| tris 2-chloroethyl phosphate, celluflex, fyrol cef, disflamoll tca, celluflex cef, niax 3cf, tris chloroethyl phosphate, trichlorethyl phosphate, 2-chloroethanol phosphate | |
| C(CCl)OP(=O)(OCCCl)OCCCl | |
| 285.482 | |
| CHEBI:35037 | |
| ≥93.0% (GC) | |
| Tris(2-chloroethyl) Phosphate |
| Yellow | |
| C6H12Cl3O4P | |
| 25 g | |
| HQUQLFOMPYWACS-UHFFFAOYSA-N | |
| tris(2-chloroethyl) phosphate | |
| 8295 | |
| 285.48 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1941
RTECSNumber : KK2450000
TSCA : Yes