missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-chloroethyl) Phosphate 93.0+%, TCI America™

Catalog No. T344725G
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T344725G 25 g
T3447100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T344725G Supplier TCI America Supplier No. T344725G
Only null left

Chemical Identifiers

CAS 115-96-8
Molecular Formula C6H12Cl3O4P
Molecular Weight (g/mol) 285.482
MDL Number MFCD00000967
InChI Key HQUQLFOMPYWACS-UHFFFAOYSA-N
Synonym tris 2-chloroethyl phosphate, celluflex, fyrol cef, disflamoll tca, celluflex cef, niax 3cf, tris chloroethyl phosphate, trichlorethyl phosphate, 2-chloroethanol phosphate
PubChem CID 8295
ChEBI CHEBI:35037
IUPAC Name tris(2-chloroethyl) phosphate
SMILES C(CCl)OP(=O)(OCCCl)OCCCl

Specifications

Color Yellow
Boiling Point 146°C
Quantity 25 g
Formula Weight 285.48
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(2-chloroethyl) Phosphate
EINECSNumber (2)-1941
RTECSNumber KK2450000
TSCA Yes