missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-carboxyethyl) Isocyanurate 98.0+%, TCI America™

Catalog No. p-7136390
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T067525G 25 g
T0675500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T067525G Supplier TCI America Supplier No. T067525G

Chemical Identifiers

CAS 2904-41-8
Molecular Formula C12H15N3O9
Molecular Weight (g/mol) 345.26
MDL Number MFCD00059790
InChI Key HENCHDCLZDQGIQ-UHFFFAOYSA-N
Synonym Isocyanuric Acid Tris(2-carboxyethyl) Ester
PubChem CID 73121
IUPAC Name 3-[3,5-bis(2-carboxyethyl)-2,4,6-trioxo-1,3,5-triazinan-1-yl]propanoic acid
SMILES OC(=O)CCN1C(=O)N(CCC(O)=O)C(=O)N(CCC(O)=O)C1=O

Specifications

Melting Point 228°C
Color White
Quantity 25 g
Formula Weight 345.26
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-carboxyethyl) Isocyanurate
EINECSNumber (5)-1062
TSCA Yes