accessibility menu, dialog, popup

UserName

HOONONOHOONOHOO

CAS RN 2904-41-8

IUPAC Name: 3-[3,5-bis(2-carboxyethyl)-2,4,6-trioxo-1,3,5-triazinan-1-yl]propanoic acid
Synonyms: 3-[3,5-bis(2-carboxyethyl)-2,4,6-trioxo-1,3,5-triazinan-1-yl]propanoic acid
Molecular Weight (g/mol): 345.26
Molecular Formula: C12H15N3O9
InChi Key: HENCHDCLZDQGIQ-UHFFFAOYSA-N
SMILES: OC(=O)CCN1C(=O)N(CCC(O)=O)C(=O)N(CCC(O)=O)C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results