missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-butoxyethyl) Phosphate 95.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 220°C |
| Quantity | 25 g |
| Formula Weight | 398.48 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | Tris(2-butoxyethyl) Phosphate |
Safety and Handling
| EINECSNumber | (2)-2022 |
| RTECSNumber | KJ9800000 |
| TSCA | Yes |