missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-butoxyethyl) phosphate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 157241000

Quantity 100g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC157241000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

78-51-3
398.48
WTLBZVNBAKMVDP-UHFFFAOYSA-N
6540
tris(2-butoxyethyl) phosphate
C18H39O7P
MFCD00009456
tris 2-butoxyethyl phosphate, tbep, phosflex t-bep, tris butoxyethyl phosphate, tributoxyethyl phosphate, tri butoxyethyl phosphate, kronitex kp-140, tributyl cellosolve phosphate, ethanol, 2-butoxy-, phosphate 3:1, tri 2-butoxyethyl phosphate
CHEBI:35038
CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC

Specifications

Tris(2-butoxyethyl) phosphate
1.0000g/mL
132°C
94% min. (GC)
C18H39O7P
[CH3(CH2)3OCH2CH2O]3P(O)
MFCD00009456
tris 2-butoxyethyl phosphate, tbep, phosflex t-bep, tris butoxyethyl phosphate, tributoxyethyl phosphate, tri butoxyethyl phosphate, kronitex kp-140, tributyl cellosolve phosphate, ethanol, 2-butoxy-, phosphate 3:1, tri 2-butoxyethyl phosphate
WTLBZVNBAKMVDP-UHFFFAOYSA-N
tris(2-butoxyethyl) phosphate
6540
398.47
78-51-3
215°C to 228°C (4.0mmHg)
Authentic
Glass bottle
1.4370 to 1.439
100g
1
Solubility in water: 1.3g/L (20°C)
CCCCOCCOP(=O)(OCCOCCCC)OCCOCCCC
398.48
CHEBI:35038
95%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-122-9

SDS
Documents

Documents

Promotions

Promotions