missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-butoxyethyl) phosphate, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111015548
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015548 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015548 Supplier Merck Emd Millipore Supplier No. 144821ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Environmental

Specifications

CAS 78-51-3
Vapor Pressure 0.03 mmHg (150°C)
Vapor Density 13.7 (vs. air)
Density 1.006 g/mL (at 25°C)
Linear Formula [CH3(CH2)3 OCH2 CH2 O]3 P(O)
MDL Number MFCD00009456
Boiling Point 215°C to 228°C (4 mmHg)
Synonym Phosphoric acid tris(2-butoxyethyl) ester
Type Analytical Standard
Quantity 1 mL
Formula Weight 398.47
Percent Purity ≥95.0% (GC)
Grade Analytical Standard
Show More Show Less