missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-benzimidazolylmethyl)amine 96.0+%, TCI America™

Supplier:  TCI America T31701G

 View more versions of this product

Catalog No. T31701G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tris(2-benzimidazolylmethyl)amine
272°C
C24H21N7
1 g
YQIGEJHOYBUSLR-UHFFFAOYSA-N
1-(1H-benzimidazol-2-yl)-N,N-bis(1H-benzimidazol-2-ylmethyl)methanamine
623893
≥96.0% (HPLC,T)
64019-57-4
White
MFCD00075516
(BimH)3
C1=CC=C2C(=C1)NC(=N2)CN(CC3=NC4=CC=CC=C4N3)CC5=NC6=CC=CC=C6N5
407.481
407.48
Crystalline Powder

Chemical Identifiers

64019-57-4
407.481
YQIGEJHOYBUSLR-UHFFFAOYSA-N
623893
C1=CC=C2C(=C1)NC(=N2)CN(CC3=NC4=CC=CC=C4N3)CC5=NC6=CC=CC=C6N5
C24H21N7
MFCD00075516
(BimH)3
1-(1H-benzimidazol-2-yl)-N,N-bis(1H-benzimidazol-2-ylmethyl)methanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions