missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-benzimidazolylmethyl)amine 96.0+%, TCI America™

Catalog No. p-7137619
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3170200MG 200 mg
T31701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3170200MG Supplier TCI America Supplier No. T3170200MG
Only null left

Chemical Identifiers

CAS 64019-57-4
Molecular Formula C24H21N7
Molecular Weight (g/mol) 407.481
MDL Number MFCD00075516
InChI Key YQIGEJHOYBUSLR-UHFFFAOYSA-N
Synonym (BimH)3
PubChem CID 623893
IUPAC Name 1-(1H-benzimidazol-2-yl)-N,N-bis(1H-benzimidazol-2-ylmethyl)methanamine
SMILES C1=CC=C2C(=C1)NC(=N2)CN(CC3=NC4=CC=CC=C4N3)CC5=NC6=CC=CC=C6N5

Specifications

Melting Point 272°C
Color White
Quantity 200 mg
Formula Weight 407.48
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2-benzimidazolylmethyl)amine