missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-aminoethyl)amine 98.0+%, TCI America™
Supplier: TCI America T12435ML
Specifications
| Tris(2-aminoethyl)amine | |
| Yellow | |
| C6H18N4 | |
| 5 mL | |
| tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine | |
| C(CN(CCN)CCN)N | |
| 146.238 | |
| CHEBI:30631 | |
| ≥98.0% (T) |
| 4097-89-6 | |
| 114°C | |
| MFCD00008177 | |
| 2922 | |
| MBYLVOKEDDQJDY-UHFFFAOYSA-N | |
| N',N'-bis(2-aminoethyl)ethane-1,2-diamine | |
| 77731 | |
| 146.24 | |
| Liquid |
Chemical Identifiers
| 4097-89-6 | |
| 146.238 | |
| MBYLVOKEDDQJDY-UHFFFAOYSA-N | |
| 77731 | |
| N',N'-bis(2-aminoethyl)ethane-1,2-diamine |
| C6H18N4 | |
| MFCD00008177 | |
| tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine | |
| CHEBI:30631 | |
| C(CN(CCN)CCN)N |
Safety and Handling
RTECSNumber : KH8587082
TSCA : Yes