missing translation for 'onlineSavingsMsg'
Learn More

Tris(2-aminoethyl)amine 98.0+%, TCI America™

Catalog No. p-7136678
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12435ML 5 mL
T124325ML 25 mL
T1243100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T12435ML Supplier TCI America Supplier No. T12435ML
Only null left

Chemical Identifiers

CAS 4097-89-6
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.238
MDL Number MFCD00008177
InChI Key MBYLVOKEDDQJDY-UHFFFAOYSA-N
Synonym tris 2-aminoethyl amine, tren, taea, 2,2',2-triaminotriethylamine, 1,2-ethanediamine, n,n-bis 2-aminoethyl, tri 2-aminoethyl amine, tren hp, tris aminoethyl amine, nitrilotris ethylamine, n,n-bis 2-aminoethyl ethane-1,2-diamine
PubChem CID 77731
ChEBI CHEBI:30631
IUPAC Name N',N'-bis(2-aminoethyl)ethane-1,2-diamine
SMILES C(CN(CCN)CCN)N

Specifications

Color Yellow
Boiling Point 114°C
Quantity 5 mL
UN Number 2922
Formula Weight 146.24
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material Tris(2-aminoethyl)amine
RTECSNumber KH8587082
TSCA Yes