missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2-acryloyloxyethyl) Isocyanurate (stabilized with Phenothiazine) 80.0+%, TCI America™
Specifications
Specifications
| Melting Point | 53°C |
| Color | White |
| Quantity | 25 g |
| UN Number | 3077 |
| Formula Weight | 423.38 |
| Percent Purity | ≥80.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | Tris(2-acryloyloxyethyl) Isocyanurate (stabilized with Phenothiazine) |
Safety and Handling
| EINECSNumber | (5)-1060 |
| TSCA | Yes |