accessibility menu, dialog, popup

UserName

CAS RN 40220-08-4

CH2OONONOOH2CONOOCH2O

CAS RN 40220-08-4

IUPAC Name: 2-{2,4,6-trioxo-3,5-bis[2-(prop-2-enoyloxy)ethyl]-1,3,5-triazinan-1-yl}ethyl prop-2-enoate
Synonyms: 2-{2,4,6-trioxo-3,5-bis[2-(prop-2-enoyloxy)ethyl]-1,3,5-triazinan-1-yl}ethyl prop-2-enoate
Molecular Weight (g/mol): 423.38
Molecular Formula: C18H21N3O9
InChi Key: YIJYFLXQHDOQGW-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCN1C(=O)N(CCOC(=O)C=C)C(=O)N(CCOC(=O)C=C)C1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results