missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)manganese(III) 98.0+%, TCI America™

Catalog No. p-7131947
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M004325G 25 g
M0043100G 100 g
M0043500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M004325G Supplier TCI America Supplier No. M004325G

Chemical Identifiers

CAS 14284-89-0
Molecular Formula C15H21MnO6
Molecular Weight (g/mol) 352.27
MDL Number MFCD00000023
InChI Key HYZQBNDRDQEWAN-LNTINUHCSA-K
Synonym mn iii acetylacetonate, tris acetylacetonato manganese iii
PubChem CID 122130696
IUPAC Name manganese(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 161°C
Color Black
Quantity 25 g
Formula Weight 352.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)manganese(III)
EINECSNumber (2)-2142
TSCA Yes