accessibility menu, dialog, popup

UserName

OOOOOOH3CCH3CH3H3CH3CH3CMn

CAS RN 14284-89-0

IUPAC Name: manganese(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Synonyms: manganese(3+) tris((2Z)-4-oxopent-2-en-2-olate)
Molecular Weight (g/mol): 352.27
Molecular Formula: C15H21MnO6
InChi Key: HYZQBNDRDQEWAN-LNTINUHCSA-K
SMILES: [Mn+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O
Physical Form 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (6)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)