missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2,4-pentanedionato)chromium(III) 98.0+%, TCI America™
Supplier: TCI America C0336100G
Specifications
| Tris(2,4-pentanedionato)chromium(III) | |
| 216°C | |
| C15H21CrO6 | |
| 100 g | |
| JWORPXLMBPOPPU-LNTINUHCSA-K | |
| chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate) | |
| 91759531 | |
| ≥98.0% (T) |
| 21679-31-2 | |
| Purple | |
| MFCD00000015 MFCD00000015 | |
| chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863 | |
| [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
| 349.32 | |
| 349.32 | |
| Crystalline Powder |
Chemical Identifiers
| 21679-31-2 | |
| 349.32 | |
| JWORPXLMBPOPPU-LNTINUHCSA-K | |
| 91759531 | |
| [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
| C15H21CrO6 | |
| MFCD00000015 MFCD00000015 | |
| chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863 | |
| chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate) |
Safety and Handling
EINECSNumber : (2)-2142
RTECSNumber : GB7575000
TSCA : Yes