missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)chromium(III) 98.0+%, TCI America™

Catalog No. p-7124361
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C033625G 25 g
C0336100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C033625G Supplier TCI America Supplier No. C033625G
Only null left

Chemical Identifiers

CAS 21679-31-2
Molecular Formula C15H21CrO6
Molecular Weight (g/mol) 349.32
MDL Number MFCD00000015 MFCD00000015
InChI Key JWORPXLMBPOPPU-LNTINUHCSA-K
Synonym chromium iii acetylacetonate, dsstox_cid_30421, dsstox_gsid_51863
PubChem CID 91759531
IUPAC Name chromium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Cr+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 216°C
Color Purple
Quantity 25 g
Formula Weight 349.32
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)chromium(III)
EINECSNumber (2)-2142
RTECSNumber GB7575000
TSCA Yes