missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tris(2,4-pentanedionato)aluminum(III) 99.0+%, TCI America™
Supplier: TCI America A0241500G
Specifications
| Tris(2,4-pentanedionato)aluminum(III) | |
| 195°C | |
| 316°C | |
| MFCD00000013 | |
| 3467 | |
| KILURZWTCGSYRE-LNTINUHCSA-K | |
| aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate) | |
| 324.31 | |
| Crystalline Powder |
| 13963-57-0 | |
| White-Yellow | |
| C15H21AlO6 | |
| 500 g | |
| Acetylacetone Aluminum(III) Salt, Aluminum(III) Acetylacetonate | |
| [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O | |
| 324.31 | |
| ≥99.0% (T) |
Chemical Identifiers
| 13963-57-0 | |
| 324.31 | |
| KILURZWTCGSYRE-LNTINUHCSA-K | |
| aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate) |
| C15H21AlO6 | |
| MFCD00000013 | |
| Acetylacetone Aluminum(III) Salt, Aluminum(III) Acetylacetonate | |
| [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O |
Safety and Handling
EINECSNumber : (2)-2142&(2)-2220
RTECSNumber : BD2230000
TSCA : Yes