missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,4-pentanedionato)aluminum(III) 99.0+%, TCI America™

Catalog No. A024125G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A024125G 25 g
A0241100G 100 g
A0241500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A024125G Supplier TCI America Supplier No. A024125G

Chemical Identifiers

CAS 13963-57-0
Molecular Formula C15H21AlO6
Molecular Weight (g/mol) 324.31
MDL Number MFCD00000013
InChI Key KILURZWTCGSYRE-LNTINUHCSA-K
Synonym Acetylacetone Aluminum(III) Salt, Aluminum(III) Acetylacetonate
IUPAC Name aluminium(3+) tris((2Z)-4-oxopent-2-en-2-olate)
SMILES [Al+3].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 195°C
Color White-Yellow
Boiling Point 316°C
Quantity 25 g
UN Number 3467
Formula Weight 324.31
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,4-pentanedionato)aluminum(III)
EINECSNumber (2)-2142&(2)-2220
RTECSNumber BD2230000
TSCA Yes