missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2'-bipyridine)cobalt(III) Tris(hexafluorophosphate) 93.0+%, TCI America™

Catalog No. T3256200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3256200MG 200 mg
T32561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3256200MG Supplier TCI America Supplier No. T3256200MG
Only null left

Chemical Identifiers

CAS 28277-53-4
Molecular Formula C30H24CoF18N6P3
Molecular Weight (g/mol) 962.39
InChI Key COCURPBAGSBJNL-UHFFFAOYSA-N
PubChem CID 15443683
IUPAC Name cobalt(3+);2-pyridin-2-ylpyridine;trihexafluorophosphate
SMILES C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.C1=CC=NC(=C1)C2=CC=CC=N2.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.[Co+3]

Specifications

Color Yellow
Quantity 200 mg
Formula Weight 962.39
Percent Purity ≥93.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,2′-bipyridine)cobalt(III) Tris(hexafluorophosphate)