accessibility menu, dialog, popup

UserName

CAS RN 28277-53-4

NNNNNNFPFFFFFFPFFFFFFPFFFFFCo

CAS RN 28277-53-4

IUPAC Name: cobalt(3+) tris(2,2'-bipyridine) tris(hexafluoro-λ⁵-phosphanuide)
Synonyms: cobalt(3+) tris(bipyridyl) trihexafluorophosphate
Molecular Weight (g/mol): 962.39
Molecular Formula: C30H24CoF18N6P3
InChi Key: COCURPBAGSBJNL-UHFFFAOYSA-N
SMILES: [Co+3].F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.F[P-](F)(F)(F)(F)F.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1.C1=CC=C(N=C1)C1=CC=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results