missing translation for 'onlineSavingsMsg'
Learn More

Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium(III) 98.0+%, TCI America™

Catalog No. T12641G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T12641G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T12641G Supplier TCI America Supplier No. T12641G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 15492-48-5
Molecular Formula C33H57O6Pr
Molecular Weight (g/mol) 690.72
MDL Number MFCD00000027,MFCD00000027
InChI Key HDONDLBRVFKILD-UHFFFAOYSA-K
Synonym pr dpm 3, praseodymium iii tris 2,2,6,6-tetramethyl-3,5-heptanedionate
PubChem CID 53384607
IUPAC Name praseodymium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
SMILES [Pr+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C

Specifications

Melting Point 220°C
Quantity 1 g
Formula Weight 690.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)praseodymium(III) [NMR Shift Reagent]