accessibility menu, dialog, popup

UserName

CAS RN 15492-48-5

CH3H3CCH3OCH3H3CCH3OCH3H3CCH3OCH3H3CCH3OCH3H3CCH3OCH3H3CCH3OPr(E/Z)(E/Z)(E/Z)

CAS RN 15492-48-5

IUPAC Name: praseodymium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Synonyms: praseodymium(3+) tris(2,2,6,6-tetramethyl-5-oxohept-3-en-3-olate)
Molecular Weight (g/mol): 690.72
Molecular Formula: C33H57O6Pr
InChi Key: HDONDLBRVFKILD-UHFFFAOYSA-K
SMILES: [Pr+3].CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C.CC(C)(C)C([O-])=CC(=O)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results