missing translation for 'onlineSavingsMsg'
Learn More

Tris(1H,1H,5H-octafluoropentyl) Phosphate 95.0+%, TCI America™

Catalog No. P113410G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P113410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P113410G Supplier TCI America Supplier No. P113410G
Only null left

Chemical Identifiers

CAS 355-86-2
Molecular Formula C15H9F24O4P
Molecular Weight (g/mol) 740.168
MDL Number MFCD00054654
InChI Key BSOLVVCARHZLMT-UHFFFAOYSA-N
Synonym Phosphoric Acid Tris(1H,1H,5H-octafluoropentyl) Ester
PubChem CID 2778037
IUPAC Name tris(2,2,3,3,4,4,5,5-octafluoropentyl) phosphate
SMILES C(C(C(C(C(F)F)(F)F)(F)F)(F)F)OP(=O)(OCC(C(C(C(F)F)(F)F)(F)F)(F)F)OCC(C(C(C(F)F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 154°C
Quantity 10 g
Formula Weight 740.17
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Tris(1H,1H,5H-octafluoropentyl) Phosphate