CAS RN 355-86-2
IUPAC Name:
tris(2,2,3,3,4,4,5,5-octafluoropentyl)phosphane
Synonyms:
tris(2,2,3,3,4,4,5,5-octafluoropentyl)phosphane
Molecular Weight (g/mol):
676.17
Molecular Formula:
C15H9F24P
InChi Key:
DISHTLVXZWCKIV-UHFFFAOYSA-N
SMILES:
FC(F)C(F)(F)C(F)(F)C(F)(F)CP(CC(F)(F)C(F)(F)C(F)(F)C(F)F)CC(F)(F)C(F)(F)C(F)(F)C(F)F
1
–
1
of
1
results
1
Tris(1H,1H,5H-octafluoropentyl) Phosphate 95.0+%, TCI America™
CAS: 355-86-2 Molecular Formula: C15H9F24O4P Molecular Weight (g/mol): 740.168 MDL Number: MFCD00054654 InChI Key: BSOLVVCARHZLMT-UHFFFAOYSA-N Synonym: Phosphoric Acid Tris(1H,1H,5H-octafluoropentyl) Ester PubChem CID: 2778037 IUPAC Name: tris(2,2,3,3,4,4,5,5-octafluoropentyl) phosphate SMILES: C(C(C(C(C(F)F)(F)F)(F)F)(F)F)OP(=O)(OCC(C(C(C(F)F)(F)F)(F)F)(F)F)OCC(C(C(C(F)F)(F)F)(F)F)(F)F